2-Bromo-9,9'-spirobi[9H-fluorene] 98%

Supplier: Ambeed
Warning

A360377-1G A360377-5G A360377-10G A360377-100G
103826-252EA 5 USD
103826-252 103837-260 76840-462 76840-464 77003-344 77881-982
2-Bromo-9,9'-spirobi[9H-fluorene] 98%
2-Bromo-9,9'-spirobi[9H-fluorene]

Precaución: for Research Use Only

Formula: C₂₅H₁₅Br
Boiling Pt: 517.2 °C (760 mm Hg)
Melting Pt: 184 °C
Flash Pt: 259.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08704217
CAS Number: 171408-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR