2-Bromo-9,9-dimethylfluorene 98%

Supplier: Ambeed
Warning

A107738-100G A107738-25G A107738-1G A107738-5G A107738-500G
103797-242EA 8 USD
103797-242 103823-010 76995-024 76995-026 77283-638 77283-640 77880-871
2-Bromo-9,9-dimethylfluorene 98%
2-Bromo-9,9-dimethylfluorene

Precaución: for Research Use Only

Formula: C₁₅H₁₃Br
MW: 273.17 g/mol
Boiling Pt: 352.9 °C (760 mm Hg)
Melting Pt: 68 °C
Flash Pt: 165.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09261272
CAS Number: 28320-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR