2-Bromo-9,10-bis(2-naphthalenyl)anthracene 97%

Supplier: Ambeed
Warning

A333420-250MG A333420-1G
103825-644EA 7 USD
103825-644 103836-610 76693-926 77306-452
2-Bromo-9,10-bis(2-naphthalenyl)anthracene 97%
2-Bromo-9,10-bis(2-naphthalenyl)anthracene

Precaución: for Research Use Only

Formula: C₃₄H₂₁Br
Boiling Pt: 630.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11045064
CAS Number: 474688-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR