2-Bromo-7-chlorobenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A184646-100MG A184646-250MG A184646-5G
103815-768EA 326 USD
103815-768 77029-592 77029-594 77871-919
2-Bromo-7-chlorobenzo[d]thiazole 95%
2-Bromo-7-chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
Storage Temperature: Refrigerator
MDL Number: MFCD09749225
CAS Number: 3507-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR