2-Bromo-6-tert-butylpyridine 95%

Supplier: Ambeed
Warning

A343414-100MG A343414-250MG
103825-884EA 64 USD
103825-884 103836-860 77263-798 77263-800 77616-508
2-Bromo-6-tert-butylpyridine 95%
2-Bromo-6-tert-butylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
Boiling Pt: 223.2±20.0 °C (760 mm Hg)
Flash Pt: 88.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607717
CAS Number: 195044-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR