2-Bromo-6-nitrobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A354082-1G
A354082-25G
A354082-5G
A354082-100G
A354082-100MG
A354082-10G
A354082-250MG
103826-114EA
45
USD
103826-114
103837-118
76840-236
76840-238
76840-240
77263-894
77263-896
77263-898
77263-900
2-Bromo-6-nitrobenzaldehyde 98%
2-Bromo-6-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₃ Boiling Pt: 320 °C (760 mm Hg) Melting Pt: 86…87 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09040530 CAS Number: 20357-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...