2-Bromo-6-nitroaniline 95%

Supplier: Ambeed
Warning

A203705-1G A203705-5G A203705-10G A203705-250MG
103817-480EA 24 USD
103817-480 103848-140 77086-274 77086-276 77260-858 77260-860
2-Bromo-6-nitroaniline 95%
2-Bromo-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
Boiling Pt: 309.8 °C (760 mm Hg)
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD09263476
CAS Number: 59255-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR