2-Bromo-6-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-picoline, 2-Bromo-6-picoline

A173929-25G A173929-100G A173929-10G A173929-500G A173929-5G
103845-184EA 5 USD
103845-184 77082-470 77273-808 77273-810 77273-812 77301-298
2-Bromo-6-methylpyridine 98%
2-Bromo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 199.5 °C (760 mm Hg)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00040743
CAS Number: 5315-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR