2-Bromo-3-hydroxy-6-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-hydroxy-2-picoline, 2-Bromo-6-methylpyridin-3-ol

A256030-1G A256030-5G
103812-232EA 23 USD
103812-232 103852-092 77091-564 77091-566
2-Bromo-3-hydroxy-6-methylpyridine 97%
2-Bromo-3-hydroxy-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.022 g/mol
Boiling Pt: 311.8 °C (760 mm Hg)
Melting Pt: 186…187 °C
Storage Temperature: Ambient
MDL Number: MFCD06655956
CAS Number: 23003-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR