2-Bromo-6-methylpyrazine 97%

Supplier: Ambeed
Warning

A302470-100MG A302470-250MG A302470-10G
103814-340EA 45 USD
103814-340 103835-498 77263-036 77263-038 77616-312
2-Bromo-6-methylpyrazine 97%
2-Bromo-6-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
Boiling Pt: 196.3 °C (760 mm Hg)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08705760
CAS Number: 914452-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR