6-Bromo-o-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methylphenol

A672373-250MG A672373-1G A672373-5G
103840-502EA 25 USD
103840-502 77035-624 77308-090 77308-092
6-Bromo-o-cresol 98%
6-Bromo-o-cresol

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.0300 g/mol
Boiling Pt: 207.9±20.0 °C (760 mm Hg)
Melting Pt: 16 °C
Flash Pt: 79.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09907876
CAS Number: 13319-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR