6-Bromo-o-toluic acid 97%

Supplier: Ambeed
Warning

A211024-1G A211024-25G A211024-5G A211024-100G A211024-10G A211024-250MG A211024-500G
103818-066EA 18 USD
103818-066 103848-758 77087-156 77087-158 77087-160 77261-054 77261-056 77261-058 77261-060
6-Bromo-o-toluic acid 97%
6-Bromo-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
Boiling Pt: 307.1 °C (760 mm Hg)
Melting Pt: 108…112 °C
Flash Pt: 139.495 °C
Storage Temperature: Ambient
MDL Number: MFCD01310788
CAS Number: 90259-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR