6-Bromo-o-toluidine 98%

Supplier: Ambeed
Warning

A169219-10G A169219-1G A169219-25G A169219-5G A169219-100G
103806-366EA 6 USD
103806-366 103844-620 76769-172 77081-692 77081-694 77081-696 77625-728
6-Bromo-o-toluidine 98%
6-Bromo-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈BrN
Boiling Pt: 243.6 °C (760 mm Hg)
Flash Pt: 101.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04039885
CAS Number: 53848-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR