2-Bromo-6-methoxypyridine 98%

Supplier: Ambeed
Warning

A111642-100G A111642-10G A111642-25G A111642-500G A111642-1G A111642-5G
103797-876EA 5 USD
103797-876 103823-694 76970-830 76970-832 76970-834 76970-836 77310-508 77310-510
2-Bromo-6-methoxypyridine 98%
2-Bromo-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 205.9 °C (760 mm Hg)
Flash Pt: 104.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00088345
CAS Number: 40473-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR