4-Amino-2-bromo-6-methoxypyridine ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-methoxypyridin-4-amine

A175175-100MG A175175-250MG
103807-004EA 87 USD
103807-004 103845-310 77163-686 77163-688
4-Amino-2-bromo-6-methoxypyridine ≥95%
4-Amino-2-bromo-6-methoxypyridine

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₆H₇BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11036355
CAS Number: 1196152-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR