2-Bromo-6-methoxyphenol 98%

Supplier: Ambeed
Warning

A118915-100G A118915-250MG A118915-25G A118915-500G A118915-1G A118915-5G
103799-134EA 6 USD
103799-134 76995-674 76995-676 76995-678 76995-680 77297-644 77311-802
2-Bromo-6-methoxyphenol 98%
2-Bromo-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
Boiling Pt: 219.4 °C (760 mm Hg)
Melting Pt: 62…65 °C
Flash Pt: 86.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08146628
CAS Number: 28165-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR