2-Bromo-6-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A154374-100G A154374-100MG A154374-10G A154374-1G A154374-25G A154374-5G
103804-630EA 11 USD
103804-630 103842-818 77259-398 77259-400 77259-402 77318-528 77318-530 77318-532
2-Bromo-6-methoxybenzoic acid 98%
2-Bromo-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Flash Pt: 144.379 °C
Storage Temperature: Ambient
MDL Number: MFCD00800732
CAS Number: 31786-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR