2-Bromo-6-methoxyaniline 97%

Supplier: Ambeed
Warning

A237282-10G A237282-250MG A237282-1G A237282-5G
103811-144EA 17 USD
103811-144 103850-922 77031-210 77031-212 77090-000 77090-002
2-Bromo-6-methoxyaniline 97%
2-Bromo-6-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.0500 g/mol
Boiling Pt: 251.3 °C (760 mm Hg)
Flash Pt: 105.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741448
CAS Number: 5473-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR