2-Bromo-6-iodo-3-methoxypyridine 97%

Supplier: Ambeed
Warning

A105494-10G A105494-250MG A105494-1G A105494-5G
103796-892EA 17 USD
103796-892 103822-624 76994-866 76994-868 77283-362 77309-492
2-Bromo-6-iodo-3-methoxypyridine 97%
2-Bromo-6-iodo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrINO
Boiling Pt: 315.7 °C (760 mm Hg)
Melting Pt: 107…109 °C
Flash Pt: 144.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909341
CAS Number: 321535-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR