6-Bromo-2-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-pyridone, 2-Bromo-6-hydroxypyridine, 6-Bromo-2(1H)-pyridinone, 6-Bromo-2-pyridinol

A156411-100G A156411-10G A156411-250MG A156411-1G A156411-25G A156411-5G
103804-876EA 9 USD
103804-876 103843-076 76997-546 76997-548 76997-550 77318-850 77318-852 77318-854
6-Bromo-2-hydroxypyridine 98%
6-Bromo-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄BrNO
MW: 174 g/mol
Flash Pt: 157.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00234043
CAS Number: 27992-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR