2-Bromo-6-fluorophenol 98%
Supplier: Ambeed
|
Warning
|
A115265-100G
A115265-10G
A115265-25G
A115265-1G
A115265-5G
103798-536EA
5
USD
103798-536
103824-390
77284-466
77284-468
77284-470
77297-356
77311-200
2-Bromo-6-fluorophenol 98%
2-Bromo-6-fluorophenol
Precaución: for Research Use Only
Formula:
C₆H₄BrFO MW: 191.0000 g/mol Boiling Pt: 173 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03788533 CAS Number: 2040-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...