2-Bromo-6-fluoronaphthalene 98%

Supplier: Ambeed
Warning

A126199-10G A126199-1G A126199-5G
103800-774EA 53 USD
103800-774 77108-866 77312-960 77312-962
2-Bromo-6-fluoronaphthalene 98%
2-Bromo-6-fluoronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrF
MW: 225.0600 g/mol
Boiling Pt: 286.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09763683
CAS Number: 324-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR