2-Bromo-6-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A401580-1G A401580-25G A401580-5G A401580-100G A401580-10G
103827-060EA 9 USD
103827-060 103838-098 76841-560 76841-562 76841-564 77278-730 77278-732
2-Bromo-6-fluorobenzoic acid 98%
2-Bromo-6-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Boiling Pt: 286.1 °C (760 mm Hg)
Melting Pt: 153…155 °C
Flash Pt: 126.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01569539
CAS Number: 2252-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR