2-Bromo-6-fluoroaniline 98%

Supplier: Ambeed
Warning

A271168-25G A271168-5G A271168-100G A271168-10G A271168-500G A271168-1G
103813-040EA 5 USD
103813-040 103852-950 77092-728 77092-730 77289-446 77289-448 77289-450 77305-068
2-Bromo-6-fluoroaniline 98%
2-Bromo-6-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 81.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01310982
CAS Number: 65896-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR