2-Bromo-6-fluoro-4-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A137126-10G
A137126-250MG
A137126-1G
A137126-25G
A137126-5G
103802-412EA
38
USD
103802-412
77258-700
77258-702
77298-890
77298-892
77298-894
2-Bromo-6-fluoro-4-methylpyridine 98%
2-Bromo-6-fluoro-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN Boiling Pt: 230.8±35.0 °C (760 mm Hg) Flash Pt: 93.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09839269 CAS Number: 180608-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...