2-Bromo-6-fluoro-3-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A126591-250MG A126591-1G
103800-840EA 29 USD
103800-840 76810-946 77298-140
2-Bromo-6-fluoro-3-methylbenzaldehyde 97%
2-Bromo-6-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Boiling Pt: 248.1 °C (760 mm Hg)
Flash Pt: 103.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11520666
CAS Number: 154650-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR