2-Bromo-6-chloropyridine 98%
Supplier: Ambeed
|
Warning
|
A394757-1G
A394757-25G
A394757-5G
A394757-100G
A394757-10G
103826-946EA
5
USD
103826-946
103837-984
76841-400
76841-402
76841-404
76978-990
76978-992
2-Bromo-6-chloropyridine 98%
2-Bromo-6-chloropyridine
Precaución: For research use only
Formula:
C₅H₃BrClN MW: 192.44 Boiling Pt: 230.8 °C (760 mm Hg) Melting Pt: 90…91 °C Flash Pt: 93.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00181262 CAS Number: 5140-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...