3-Amino-2-bromo-6-chloropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-6-chloro-2-bromopyridine, 2-Bromo-6-chloro-3-pyridylamine, 2-Bromo-6-chloropyridin-3-amine

A486606-1G A486606-5G A486606-100G
103828-180EA 11 USD
103828-180 103839-254 76843-196 76843-198 77669-174
3-Amino-2-bromo-6-chloropyridine 97%
3-Amino-2-bromo-6-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Boiling Pt: 330.3±37.0 °C (760 mm Hg)
Flash Pt: 153.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11504927
CAS Number: 1050501-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR