2-Bromo-6-chlorophenol 97%

Supplier: Ambeed
Warning

A197672-100G A197672-250MG A197672-1G A197672-5G
103816-954EA 11 USD
103816-954 103847-576 76873-282 76873-284 77085-592 77085-594
2-Bromo-6-chlorophenol 97%
2-Bromo-6-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
Boiling Pt: 204.6 °C (760 mm Hg)
Melting Pt: 50…54 °C
Flash Pt: 77.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09907875
CAS Number: 2040-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR