2-Bromo-6-chloro-pyridine-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A102064-250MG A102064-100MG
103796-274EA 194 USD
103796-274 103821-970 76821-276 77610-762
2-Bromo-6-chloro-pyridine-3-carbaldehyde 95%
2-Bromo-6-chloro-pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11844804
CAS Number: 1060815-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR