2-Bromo-6-chlorobenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A109229-250MG
A109229-25G
A109229-1G
A109229-5G
103797-472EA
26
USD
103797-472
103823-264
76765-682
76765-684
77310-088
77310-090
2-Bromo-6-chlorobenzonitrile 97%
2-Bromo-6-chlorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃BrClN MW: 216.46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06335184 CAS Number: 6575-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...