2-Bromo-6-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A462516-25G A462516-5G A462516-100G A462516-10G A462516-500G A462516-1G
103827-942EA 10 USD
103827-942 103839-018 76842-906 76842-908 77116-302 77116-304 77116-306 77307-434
2-Bromo-6-chlorobenzoic acid 98%
2-Bromo-6-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 148…152 °C
Flash Pt: 144.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00672929
CAS Number: 93224-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR