2-Bromo-6-chlorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A158110-10G A158110-1G A158110-5G
103805-094EA 36 USD
103805-094 103843-302 76973-408 77300-316 77300-318
2-Bromo-6-chlorobenzo[d]thiazole 97%
2-Bromo-6-chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MW: 248.5300 g/mol
Boiling Pt: 330.5 °C (760 mm Hg)
Flash Pt: 153.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783781
CAS Number: 3507-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR