2-Bromo-6-chlorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A175452-100G
A175452-10G
A175452-250MG
A175452-25G
A175452-5G
A175452-1G
103807-040EA
6
USD
103807-040
103845-348
76784-414
76784-416
76784-418
77082-690
77082-692
77301-372
2-Bromo-6-chlorobenzaldehyde 97%
2-Bromo-6-chlorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄BrClO Melting Pt: 74…76 °C Flash Pt: 110.33 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00266792 CAS Number: 64622-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...