2-Bromo-6-chloroaniline 95%

Supplier: Ambeed
Warning

A436339-1G A436339-25G A436339-5G A436339-10G A436339-250MG
103827-612EA 15 USD
103827-612 103838-676 76842-410 76842-412 76842-414 76990-576 76990-578
2-Bromo-6-chloroaniline 95%
2-Bromo-6-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 242.8 °C (760 mm Hg)
Flash Pt: 100.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977383
CAS Number: 59772-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR