2-Bromo-6-chloro-3-nitropyridine 95%

Supplier: Ambeed
Warning

A300325-10G A300325-1G A300325-25G A300325-250MG A300325-100MG
103814-270EA 21 USD
103814-270 103835-422 77262-986 77305-806 77305-808 77616-302 77616-300
2-Bromo-6-chloro-3-nitropyridine 95%
2-Bromo-6-chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
MW: 237.4400 g/mol
Boiling Pt: 277.2 °C (760 mm Hg)
Flash Pt: 121.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260904
CAS Number: 91678-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR