2-Bromo-6-chloro-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A264814-100MG A264814-250MG
103812-692EA 63 USD
103812-692 103852-586 77238-604 77238-606
2-Bromo-6-chloro-1H-benzo[d]imidazole 97%
2-Bromo-6-chloro-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22562661
CAS Number: 1388031-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR