2-Bromo-6,7-dihydrothiazolo[5,4-c]pyridin-4(5H)-one 95%

Supplier: Ambeed
Warning

A151920-250MG A151920-1G A151920-100MG
103804-318EA 611 USD
103804-318 103842-468 77028-166 77299-912 77871-749
2-Bromo-6,7-dihydrothiazolo[5,4-c]pyridin-4(5H)-one 95%
2-Bromo-6,7-dihydrothiazolo[5,4-c]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₆H₅BrN₂OS
Boiling Pt: 487.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD14706758
CAS Number: 1035219-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR