2-Bromo-6-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-carboxybenzotrifluoride, 6-Bromo-α,α,α-trifluoro-o-toluic acid

A255816-250MG A255816-1G A255816-5G
103812-216EA 41 USD
103812-216 103852-076 76815-840 77091-540 77091-542
2-Bromo-6-(trifluoromethyl)benzoic acid 98%
2-Bromo-6-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD06253757
CAS Number: 177420-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR