2-Bromo-5-phenylpyridine 97%

Supplier: Ambeed
Warning

A345314-1G A345314-5G A345314-10G
103825-920EA 21 USD
103825-920 103836-902 76839-958 76839-960 77263-804
2-Bromo-5-phenylpyridine 97%
2-Bromo-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
Boiling Pt: 335.3 °C (760 mm Hg)
Melting Pt: 75 °C
Flash Pt: 156.6 °C
Storage Temperature: Ambient
MDL Number: MFCD04114195
CAS Number: 107351-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR