2-Bromo-5-nitropyrazine 95%

Supplier: Ambeed
Warning

A357082-100MG A357082-250MG
103826-182EA 318 USD
103826-182 77033-158 77672-792
2-Bromo-5-nitropyrazine 95%
2-Bromo-5-nitropyrazine

Precaución: for Research Use Only

Formula: C₄H₂BrN₃O₂
Boiling Pt: 298.295 °C (760 mm Hg)
Melting Pt: 114 °C
Flash Pt: 134.204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11870741
CAS Number: 117103-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR