2-Bromo-5-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A110506-100G A110506-10G A110506-25G A110506-1G A110506-5G
103797-676EA 5 USD
103797-676 103823-482 76995-192 76995-194 76995-196 77310-276 77310-278
2-Bromo-5-nitrobenzaldehyde 98%
2-Bromo-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃
Flash Pt: 139.574 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00462865
CAS Number: 84459-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR