2-Bromo-5-nitroanisole 98%

Supplier: Ambeed
Warning

A182128-1G A182128-100G
103846-054EA 9 USD
103846-054 77138-760 77138-762 77746-183
2-Bromo-5-nitroanisole 98%
2-Bromo-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
Flash Pt: 136.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00041250
CAS Number: 77337-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR