2-Bromo-5-methylthiophene 98% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol
Stabilization:
Stabilized with Copper chip + NaHCO3
A100942-100G
A100942-10G
A100942-1G
A100942-25G
A100942-5G
103821-404EA
9
USD
103821-404
77282-612
77282-614
77282-616
77282-618
77296-922
2-Bromo-5-methylthiophene 98% stabilized
2-Bromo-5-methylthiophene
Precaución: for Research Use Only
Formula:
C₅H₅BrS MW: 177.06 g/mol Boiling Pt: 176.7 °C (760 mm Hg) Flash Pt: 78.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00130103 CAS Number: 765-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...