2-Bromo-5-methylpyrimidine 95%

Supplier: Ambeed
Warning

A257537-100MG A257537-250MG A257537-1G A257537-5G
103812-316EA 53 USD
103812-316 103852-184 76786-902 76786-904 77091-696 77091-698
2-Bromo-5-methylpyrimidine 95%
2-Bromo-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
Boiling Pt: 265 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09753737
CAS Number: 150010-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR