2-Bromo-5-methylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A229249-100MG A229249-250MG A229249-1G A229249-5G
103819-458EA 18 USD
103819-458 103850-358 76826-400 76826-402 77089-186 77089-188
2-Bromo-5-methylpyridin-4-amine 95%
2-Bromo-5-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
Melting Pt: 130…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100653
CAS Number: 79055-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR