2-Bromo-5-methylphenol 98%
Supplier: Ambeed
|
Warning
|
A224829-100MG
A224829-10G
A224829-250MG
A224829-1G
A224829-25G
A224829-5G
103819-134EA
8
USD
103819-134
103850-026
76874-060
76874-062
76874-064
77303-712
77303-714
77303-716
2-Bromo-5-methylphenol 98%
2-Bromo-5-methylphenol
Precaución: for Research Use Only
Formula:
C₇H₇BrO MW: 187.034 g/mol Boiling Pt: 218.2 °C (760 mm Hg) Flash Pt: 85.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11100989 CAS Number: 14847-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...