6-Bromo-m-toluic acid 97%

Supplier: Ambeed
Warning

A110542-100G A110542-10G A110542-25G A110542-5G
103797-680EA 5 USD
103797-680 103823-486 76687-522 76687-524 76687-526 77310-284
6-Bromo-m-toluic acid 97%
6-Bromo-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 136…140 °C
Flash Pt: 142.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00079722
CAS Number: 6967-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR