2-Bromo-5-methyl-4-nitropyridine 98%

Supplier: Ambeed
Warning

A146384-100MG A146384-250MG A146384-1G
103803-660EA 29 USD
103803-660 103841-750 76972-858 76972-860 77317-074
2-Bromo-5-methyl-4-nitropyridine 98%
2-Bromo-5-methyl-4-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Flash Pt: 135.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100652
CAS Number: 66092-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR