2-Bromo-5-methyl-3-nitropyridine 97%
Supplier: Ambeed
|
Warning
|
A115338-100G
A115338-25G
A115338-1G
A115338-5G
103798-550EA
9
USD
103798-550
103824-404
77136-210
77136-212
77311-218
77311-220
2-Bromo-5-methyl-3-nitropyridine 97%
2-Bromo-5-methyl-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrN₂O₂ Boiling Pt: 276.7 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08703667 CAS Number: 23056-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...